Assay Details
| Gene Symbol: Large | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010687.1 | NP_034817.1 | 13 - 14 | 2053 | 77 | |
| GenBank mRNA | AJ006278.1 | - | 13 - 14 | 2053 | 77 | |
| AK122328.1 | - | 14 - 15 | 3130 | 77 | ||
| AK133940.1 | - | 13 - 14 | 2538 | 77 | ||
| BC061506.1 | - | 12 - 13 | 1810 | 77 | ||
| BC079596.1 | - | 13 - 14 | 2334 | 77 | ||
| BC100399.1 | - | 13 - 14 | 2116 | 30725031 | 77 | |
| BC125487.1 | - | 13 - 14 | 2019 | 40130002 | 77 | |
| BC125489.1 | - | 13 - 14 | 2019 | 40130004 | 77 | |
| HM141820.1 | - | 11 - 12 | 1688 | 77 | ||
More Information
Set Membership: |
|

Large
Genomic Map