Assay Details
| Gene Symbol: Lmnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010721.2 | NP_034851.2 | 6 - 7 | 1462 | 69 | |
| GenBank mRNA | AK079511.1 | - | 6 - 7 | 1463 | 69 | |
| AK089110.1 | - | 6 - 7 | 1428 | 69 | ||
| AK132252.1 | - | 6 - 7 | 1464 | 69 | ||
| BC052729.1 | - | 6 - 7 | 1411 | 6816118 | 69 | |
| BC058392.1 | - | 6 - 7 | 1411 | 6816118 | 69 | |
| M35153.1 | - | 6 - 7 | 1417 | 69 | ||
| X16705.1 | - | 6 - 7 | 1416 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmnb1
Genomic Map