Assay Details
| Gene Symbol: Mbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010774.2 | NP_034904.2 | 6 - 7 | 1613 | 73 | |
| GenBank mRNA | AF072249.1 | - | 6 - 7 | 1630 | 73 | |
| AK147782.1 | - | 3 - 4 | 595 | 73 | ||
| AK161005.1 | - | 6 - 7 | 2601 | 73 | ||
| BC024812.1 | - | 6 - 7 | 1595 | 5356016 | 73 | |
More Information
Set Membership: |
|

Mbd4
Genomic Map