Assay Details
| Gene Symbol: Lgi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020278.2 | NP_064674.1 | 2 - 3 | 386 | 136 | |
| GenBank mRNA | AK045443.1 | - | 2 - 3 | 491 | 136 | |
| BC066090.1 | - | 2 - 3 | 386 | 6839091 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgi1
Genomic Map