Assay Details
| Gene Symbol: Slc39a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135151.1 | NP_001128623.1 | 7 - 8 | 1183 | 73 | |
| NM_001135152.1 | NP_001128624.1 | 7 - 8 | 1149 | 73 | ||
| NM_144808.4 | NP_659057.2 | 7 - 8 | 1183 | 73 | ||
| GenBank mRNA | AB177995.1 | - | 7 - 8 | 1186 | 73 | |
| AK090193.1 | - | 7 - 8 | 1146 | 73 | ||
| AK122197.1 | - | 738 | 73 | |||
| AK192726.1 | - | 2 - 3 | 250 | 73 | ||
| BC021530.1 | - | 7 - 8 | 1186 | 5353717 | 73 | |
| CB521415.1 | - | 4 - 5 | 511 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a14
Genomic Map