Assay Details
| Gene Symbol: Ncaph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144818.3 | NP_659067.2 | 9 - 10 | 1273 | 116 | |
| GenBank mRNA | AK028919.1 | - | 9 - 10 | 1258 | 116 | |
| AK042706.1 | - | 9 - 10 | 1190 | 116 | ||
| AK080433.1 | - | 9 - 10 | 1262 | 116 | ||
| AK129047.1 | - | 6 - 7 | 826 | 116 | ||
| BC021499.1 | - | 9 - 10 | 1247 | 5346298 | 116 | |
| BC025060.1 | - | 9 - 10 | 1198 | 3673890 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncaph
Genomic Map