Assay Details
| Gene Symbol: Ncaph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144818.3 | NP_659067.2 | 11 - 12 | 1528 | 128 | |
| GenBank mRNA | AK028919.1 | - | 11 - 12 | 1513 | 128 | |
| AK042706.1 | - | 11 - 12 | 1445 | 128 | ||
| AK080433.1 | - | 11 - 12 | 1517 | 128 | ||
| AK129047.1 | - | 8 - 9 | 1081 | 128 | ||
| BC021499.1 | - | 11 - 12 | 1502 | 5346298 | 128 | |
| BC025060.1 | - | 11 - 12 | 1453 | 3673890 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncaph
Genomic Map