Assay Details
| Gene Symbol: Tmem106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144830.3 | NP_659079.1 | 2 - 3 | 159 | 63 | |
| GenBank mRNA | AK146019.1 | - | 2 - 3 | 160 | 63 | |
| AK149650.1 | - | 1 - 2 | 403 | 63 | ||
| AK150285.1 | - | 2 - 3 | 129 | 63 | ||
| AK151842.1 | - | 2 - 3 | 136 | 63 | ||
| AK152741.1 | - | 2 - 3 | 129 | 63 | ||
| AK154217.1 | - | 2 - 3 | 160 | 63 | ||
| AK168437.1 | - | 2 - 3 | 102 | 63 | ||
| AK169081.1 | - | 2 - 3 | 102 | 63 | ||
| AK170900.1 | - | 2 - 3 | 130 | 63 | ||
| AK172282.1 | - | 2 - 3 | 108 | 63 | ||
| BC022145.1 | - | 1 - 2 | 207 | 5101508 | 63 | |
| BC033268.1 | - | 2 - 3 | 132 | 5037750 | 63 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK146019.1,AK149650.1,AK150285.1,AK151842.1,AK152741.1,AK154217.1,AK169081.1,AK170900.1,AK172282.1,BC022145.1,BC033268.1,NM_144830.3 transcripts.
Set Membership: |
|

Tmem106a
Genomic Map