Assay Details
| Gene Symbol: Tmem106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144830.3 | NP_659079.1 | 3 - 4 | 393 | 72 | |
| GenBank mRNA | AK146019.1 | - | 3 - 4 | 394 | 72 | |
| AK149650.1 | - | 2 - 3 | 637 | 72 | ||
| AK150285.1 | - | 3 - 4 | 363 | 72 | ||
| AK151842.1 | - | 3 - 4 | 369 | 72 | ||
| AK152741.1 | - | 3 - 4 | 363 | 72 | ||
| AK154217.1 | - | 3 - 4 | 394 | 72 | ||
| AK168437.1 | - | 3 - 4 | 336 | 72 | ||
| AK169081.1 | - | 3 - 4 | 336 | 72 | ||
| AK170900.1 | - | 3 - 4 | 364 | 72 | ||
| AK172282.1 | - | 3 - 4 | 342 | 72 | ||
| BC022145.1 | - | 2 - 3 | 441 | 5101508 | 72 | |
More Information
Set Membership: |
|

Tmem106a
Genomic Map