| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100015636.mm.1 |
NM_144830.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015636.mm.1 |
NM_144830.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015636.mm.1 |
TC1100001661.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015637.mm.1 |
NM_144830.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015637.mm.1 |
NM_144830.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015637.mm.1 |
TC1100001661.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015633.mm.1 |
NM_144830.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015633.mm.1 |
NM_144830.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015633.mm.1 |
TC1100001661.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015635.mm.1 |
NM_144830.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015635.mm.1 |
NM_144830.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015635.mm.1 |
TC1100001661.mm.1 |
MTA TC |