Assay Details
| Gene Symbol: Ugt3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144845.3 | NP_659094.1 | 1 - 2 | 194 | 83 | |
| GenBank mRNA | AK041045.1 | - | 1 - 2 | 179 | 83 | |
| AK050128.1 | - | 1 - 2 | 195 | 83 | ||
| AK052644.1 | - | 1 - 2 | 204 | 83 | ||
| AK143815.1 | - | 1 - 2 | 209 | 83 | ||
| BC022134.1 | - | 1 - 2 | 179 | 5098754 | 83 | |
| BC034837.1 | - | 1 - 2 | 175 | 5134131 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ugt3a2
Genomic Map