| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700022311.mm.1 |
NM_001025309.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022311.mm.1 |
NM_001025309.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022311.mm.1 |
NM_144859.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022311.mm.1 |
NM_144859.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022311.mm.1 |
TC1700002400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022312.mm.1 |
NM_001025309.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022312.mm.1 |
NM_001025309.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022312.mm.1 |
NM_144859.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022312.mm.1 |
NM_144859.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022312.mm.1 |
TC1700002400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022313.mm.1 |
NM_001025309.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022313.mm.1 |
NM_001025309.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022313.mm.1 |
NM_144859.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022313.mm.1 |
NM_144859.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022313.mm.1 |
TC1700002400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022314.mm.1 |
NM_001025309.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022314.mm.1 |
NM_001025309.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022314.mm.1 |
NM_144859.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022314.mm.1 |
NM_144859.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022314.mm.1 |
TC1700002400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022315.mm.1 |
NM_001025309.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022315.mm.1 |
NM_001025309.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022315.mm.1 |
NM_144859.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022315.mm.1 |
NM_144859.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022315.mm.1 |
TC1700002400.mm.1 |
MTA TC |