Assay Details
| Gene Symbol: Lims2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144862.3 | NP_659111.1 | 1 - 2 | 46 | 64 | |
| GenBank mRNA | BC010816.1 | - | 1 - 2 | 47 | 4212305 | 64 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC010816.1,NM_144862.3 transcripts.
Set Membership: |
|

Lims2
Genomic Map