Assay Details
| Gene Symbol: Lims2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144862.3 | NP_659111.1 | 6 - 7 | 696 | 63 | |
| GenBank mRNA | BC010816.1 | - | 6 - 7 | 697 | 4212305 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Lims2
Genomic Map