Assay Details
| Gene Symbol: Spg20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144987.1 | NP_001138459.1 | 5 - 6 | 1627 | 79 | |
| NM_001144988.1 | NP_001138460.1 | 4 - 5 | 1240 | 79 | ||
| NM_144895.2 | NP_659144.1 | 5 - 6 | 1411 | 79 | ||
| GenBank mRNA | AK031545.1 | - | 4 - 5 | 966 | 79 | |
| AK035081.1 | - | 3 - 4 | 1099 | 79 | ||
| AK036569.1 | - | 3 - 4 | 563 | 79 | ||
| AK087388.1 | - | 5 - 6 | 1380 | 79 | ||
| AK089946.1 | - | 5 - 6 | 1412 | 79 | ||
| AK129171.1 | - | 4 - 5 | 1219 | 79 | ||
| AK147760.1 | - | 5 - 6 | 1629 | 79 | ||
| AK159951.1 | - | 5 - 6 | 1412 | 79 | ||
| AK200807.1 | - | 2 - 3 | 147 | 79 | ||
| BC022921.1 | - | 5 - 6 | 1379 | 4220506 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spg20
Genomic Map