Assay Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144902.3 | NP_659151.1 | 3 - 4 | 555 | 120 | |
| GenBank mRNA | AK029398.1 | - | 3 - 4 | 927 | 120 | |
| AK041195.1 | - | 4 - 5 | 675 | 120 | ||
| AK049484.1 | - | 3 - 4 | 555 | 120 | ||
| AK161317.1 | - | 3 - 4 | 513 | 120 | ||
| BC024110.1 | - | 3 - 4 | 551 | 5150304 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35a3
Genomic Map