Assay Details
| Gene Symbol: Ptbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144904.2 | NP_659153.2 | 2 - 3 | 249 | 76 | |
| GenBank mRNA | BC006638.1 | - | 2 - 3 | 159 | 3969488 | 76 |
| CJ103126.1 | - | 2 - 3 | 181 | 76 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC006638.1,NM_144904.2 transcripts.
Set Membership: |
|

Ptbp3
Genomic Map