Assay Details
| Gene Symbol: Tmem150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144916.3 | NP_659165.1 | 4 - 5 | 379 | 83 | |
| GenBank mRNA | BC014685.1 | - | 3 - 4 | 295 | 4241375 | 83 |
| BC028825.1 | - | 4 - 5 | 381 | 5097962 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem150a
Genomic Map