Assay Details
| Gene Symbol: Tmem150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144916.3 | NP_659165.1 | 7 - 8 | 756 | 73 | |
| GenBank mRNA | BC014685.1 | - | 6 - 7 | 672 | 4241375 | 73 |
| BC028825.1 | - | 7 - 8 | 758 | 5097962 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541279 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541280 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P164939 | NM_144916.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P164939 | BC014685.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P164939 | BC028825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P164939 | NM_144916.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P164939 | BC014685.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P164939 | BC028825.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539701 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P164939 | NM_144916.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P164939 | BC014685.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P164939 | BC028825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P164939 | NM_144916.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P164939 | BC014685.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P164939 | BC028825.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24541284 | - | - |
| Whole Mouse Genome G4122A | A_51_P164939 | NM_144916.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P164939 | BC014685.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P164939 | BC028825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P164939 | NM_144916.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P164939 | BC014685.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P164939 | BC028825.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131718 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05619238 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05619247 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05619232 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541279 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24541280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05716367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05313603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05716375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05716370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05619238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05716379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05580947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05075283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05580939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541286 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539113 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539112 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539114 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424726_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539111 | - | - |
| Mouse Genome 430 2.0 Array | 1424726_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424726_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232086_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232086_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600006491.mm.1 | NM_144916.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006491.mm.1 | NM_144916.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006491.mm.1 | TC0600000849.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424726_at | - | - |
| Mouse Expression Set 430 | 1424726_PM_at | - | - |
| Murine Genome U74v2 Set | 103078_PM_at | - | - |
| Murine Genome U74v2 Set | 103078_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424726_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424726_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424726_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424726_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 636279 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2120605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002120605 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2653396 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2653396 | - | - |
| mouse-6 v1.1 (Illumina) | scl29884.7.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2120605 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29884.7.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2653395 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2861123 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2653395 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2861123 | - | - |

Tmem150a
Genomic Map