Assay Details
| Gene Symbol: Wasf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145155.3 | NP_660137.1 | 4 - 5 | 701 | 99 | |
| GenBank mRNA | AF454703.1 | - | 3 - 4 | 320 | 99 | |
| AK044481.1 | - | 722 | 99 | |||
| BC027038.1 | - | 4 - 5 | 701 | 5363785 | 99 | |
More Information
Set Membership: |
|

Wasf3
Genomic Map