Assay Details
| Gene Symbol: Sdcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145535.2 | NP_663510.1 | 4 - 5 | 331 | 117 | |
| GenBank mRNA | AK136572.1 | - | 4 - 5 | 287 | 117 | |
| AK144452.1 | - | 4 - 5 | 296 | 117 | ||
| AK160483.1 | - | 4 - 5 | 289 | 117 | ||
| BC005556.1 | - | 4 - 5 | 322 | 3964815 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sdcbp2
Genomic Map