Assay Details
| Gene Symbol: Clcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177770.1 | NP_001171241.1 | 5 - 6 | 931 | 94 | |
| NM_001177771.1 | NP_001171242.1 | 6 - 7 | 846 | 94 | ||
| NM_145543.2 | NP_663518.1 | 5 - 6 | 643 | 94 | ||
| GenBank mRNA | AK090092.1 | - | 5 - 6 | 617 | 95 | |
| AK143631.1 | - | 6 - 7 | 829 | 94 | ||
| AK169803.1 | - | 5 - 6 | 641 | 94 | ||
| AK171296.1 | - | 5 - 6 | 670 | 94 | ||
| BC003247.1 | - | 5 - 6 | 674 | 3498611 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcc1
Genomic Map