Assay Details
| Gene Symbol: Clcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177770.1 | NP_001171241.1 | 7 - 8 | 1160 | 103 | |
| NM_001177771.1 | NP_001171242.1 | 8 - 9 | 1075 | 103 | ||
| NM_145543.2 | NP_663518.1 | 7 - 8 | 872 | 103 | ||
| GenBank mRNA | AK090092.1 | - | 7 - 8 | 847 | 103 | |
| AK143631.1 | - | 8 - 9 | 1058 | 103 | ||
| AK169803.1 | - | 7 - 8 | 870 | 103 | ||
| AK171296.1 | - | 7 - 8 | 899 | 103 | ||
| BC003247.1 | - | 7 - 8 | 903 | 3498611 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcc1
Genomic Map