Assay Details
| Gene Symbol: Mob1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145571.2 | NP_663546.1 | 3 - 4 | 490 | 91 | |
| GenBank mRNA | AK158228.1 | - | 3 - 4 | 473 | 91 | |
| AK167515.1 | - | 3 - 4 | 485 | 91 | ||
| AK170691.1 | - | 3 - 4 | 484 | 91 | ||
| AK172205.1 | - | 3 - 4 | 493 | 91 | ||
| AK216221.1 | - | 2 - 3 | 181 | 91 | ||
| BC009149.1 | - | 3 - 4 | 490 | 2651599 | 91 | |
| BC033463.1 | - | 3 - 4 | 444 | 3500664 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mob1a
Genomic Map