Assay Details
| Gene Symbol: Ces2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145603.2 | NP_663578.1 | 5 - 6 | 882 | 78 | |
| GenBank mRNA | AB110073.1 | - | 5 - 6 | 882 | 78 | |
| BC015290.1 | - | 5 - 6 | 866 | 4241028 | 78 | |
| BC024552.1 | - | 5 - 6 | 864 | 4990820 | 78 | |
| BC031170.1 | - | 5 - 6 | 864 | 4984806 | 78 | |
| BC034178.1 | - | 5 - 6 | 851 | 4971285 | 78 | |
| BC034180.1 | - | 5 - 6 | 864 | 4972755 | 78 | |
| BC034182.1 | - | 1 - 2 | 182 | 78 | ||
| BC034191.1 | - | 5 - 6 | 864 | 4982995 | 78 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC034182.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124712 | NR_033726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124712 | AK078958.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124712 | BC034182.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124712 | BC034191.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124712 | BC031170.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | NR_033726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | AK078958.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | BC034182.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | BC034191.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | BC031170.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | NR_033726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | AK078958.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | BC034182.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | BC034191.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P318361 | BC031170.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005213 | NR_033726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005213 | AK078958.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005213 | BC034182.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005213 | BC034191.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005213 | BC031170.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42425 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157819 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | NR_033726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | AK078958.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | BC034182.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | BC034191.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | BC031170.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | NR_033726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | AK078958.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | BC034182.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | BC034191.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P318361 | BC031170.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005213 | NR_033726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005213 | AK078958.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005213 | BC034182.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005213 | BC034191.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005213 | BC031170.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124712 | NR_033726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124712 | AK078958.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124712 | BC034182.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124712 | BC034191.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124712 | BC031170.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26130525 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157819 | - | - |
| Whole Mouse Genome G4122A | A_51_P513573 | NR_033726.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513573 | BC034182.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513573 | BC034191.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513573 | BC031170.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513573 | BC034178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | NR_033726.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | AK078958.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | BC034182.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | BC034191.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | BC031170.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | NR_033726.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | AK078958.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | BC034182.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | BC034191.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318361 | BC031170.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106825 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157819 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07048223 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07048190 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07048211 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07048202 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07048207 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07048222 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07048214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26130505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07048195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06943774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07048216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06859704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07048209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06860812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26130490 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10574535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574542 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574540 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574543 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574536 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574537 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574539 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574534 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574544 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574538 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574541 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574528 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10574524 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10574532 | - | - |
| Mouse Genome 430 2.0 Array | 1424245_at | - | - |
| Mouse Genome 430 2.0 Array | 1424245_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800009287.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009287.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009287.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009296.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009296.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009296.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009297.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009297.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009297.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009290.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009290.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009290.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009292.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009292.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009292.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009289.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009289.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009289.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009301.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009301.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009301.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009294.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009294.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009294.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009295.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009295.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009295.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009288.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009288.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009288.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009293.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009293.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009293.mm.1 | TC0800001194.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009291.mm.1 | NM_145603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009291.mm.1 | NM_145603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009291.mm.1 | TC0800001194.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424245_PM_at | - | - |
| Mouse Expression Set 430 | 1424245_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424245_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424245_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 850024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5340609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5340609 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005340609 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2743848 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2743848 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5340609 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340609 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2743848 | - | - |

Ces2c
Genomic Map