Assay Details
| Gene Symbol: Ppan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145610.2 | NP_663585.2 | 1 - 2 | 149 | 61 | |
| GenBank mRNA | AK039027.1 | - | 1 - 2 | 54 | 61 | |
| BC014688.1 | - | 1 - 2 | 67 | 4489442 | 61 | |
| BC059277.1 | - | 1 - 2 | 39 | 61 | ||
| CJ077201.1 | - | 1 - 2 | 150 | 61 | ||
| Gene Symbol: B130055M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK156871.1 | - | 58 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK039027.1,BC014688.1,NM_145610.2 transcripts.
Set Membership: |
|

Ppan
Genomic Map