Assay Details
| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145939.2 | NP_666051.2 | 1 - 2 | 201 | 63 | |
| GenBank mRNA | BC031893.1 | - | 1 - 2 | 196 | 5371807 | 63 |
| CA538864.1 | - | 1 - 2 | 201 | 63 | ||
| CX217502.1 | - | 1 - 2 | 148 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30588474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30588476 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30588475 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155501 | AK090067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155501 | AK149510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155501 | NM_145939.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155501 | BC031893.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155501 | AK090067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155501 | AK149510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155501 | NM_145939.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155501 | BC031893.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563863 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889856 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159678 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155501 | AK090067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155501 | AK149510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155501 | NM_145939.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155501 | BC031893.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155501 | AK090067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155501 | AK149510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155501 | NM_145939.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155501 | BC031893.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155501 | AK090067.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155501 | AK149510.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155501 | NM_145939.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155501 | BC031893.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155501 | AK090067.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155501 | AK149510.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155501 | NM_145939.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155501 | BC031893.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588437 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159678 | - | - |
| Whole Mouse Genome G4122A | A_51_P155501 | AK090067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155501 | AK149510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155501 | NM_145939.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155501 | BC031893.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155501 | AK090067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155501 | AK149510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155501 | NM_145939.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155501 | BC031893.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138168 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129343 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159678 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12958422 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12958426 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12958427 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12958415 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12958413 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30588474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30588476 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30588475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12639049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12761435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12761400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13053808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12761409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12761424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12761415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12958424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12761404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12761407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13053801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588453 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10438524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438519 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460742_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10438517 | - | - |
| Mouse Genome 430 2.0 Array | 1460742_at | - | - |
| Mouse Genome 430 2.0 Array | 1460742_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208624_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208624_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600011201.mm.1 | NM_145939.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011201.mm.1 | NM_145939.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011201.mm.1 | TC1600001349.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600011200.mm.1 | NM_145939.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011200.mm.1 | NM_145939.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011200.mm.1 | TC1600001349.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600011202.mm.1 | NM_145939.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011202.mm.1 | NM_145939.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011202.mm.1 | TC1600001349.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460742_PM_at | - | - |
| Mouse Expression Set 430 | 1460742_at | - | - |
| Murine Genome U74v2 Set | 162337_f_at | - | - |
| Murine Genome U74v2 Set | 103790_PM_at | - | - |
| Murine Genome U74v2 Set | 162337_PM_f_at | - | - |
| Murine Genome U74v2 Set | 103790_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460742_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460742_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460742_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460742_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 458043 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7100131 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2650647 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7100131 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650647 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007100131 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2619826 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215893 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215893 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2619826 | - | - |
| mouse-6 v1.1 (Illumina) | scl48662.9.1_15-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001901.1_31-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7100131 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650647 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001901.1_31-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48662.9.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7100131 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650647 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1215893 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2744459 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2910106 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2910106 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2744459 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215893 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001901.1_31-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl48662.9.1_15-S | - | - |

Alg3
Genomic Map