Assay Details
| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145939.2 | NP_666051.2 | 4 - 5 | 615 | 89 | |
| GenBank mRNA | AK090067.1 | - | 3 - 4 | 516 | 89 | |
| BC031893.1 | - | 4 - 5 | 610 | 5371807 | 89 | |
| CX217502.1 | - | 4 - 5 | 562 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alg3
Genomic Map