Assay Details
| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145939.2 | NP_666051.2 | 6 - 7 | 945 | 64 | |
| GenBank mRNA | AK090067.1 | - | 5 - 6 | 846 | 64 | |
| AK149510.1 | - | 5 - 6 | 845 | 64 | ||
| BC031893.1 | - | 6 - 7 | 940 | 5371807 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alg3
Genomic Map