Assay Details
| Gene Symbol: Prr15l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146026.1 | NP_666138.1 | 2 - 3 | 227 | 65 | |
| GenBank mRNA | BC022765.1 | - | 2 - 3 | 227 | 4925830 | 65 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC022765.1,NM_146026.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036764 | BC036171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036764 | BC022765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036764 | BC024510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036764 | AK090284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036764 | BC034192.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | BC036171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | BC022765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | BC024510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | AK090284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | BC034192.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | BC036171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | BC022765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | BC024510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | AK090284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P435561 | BC034192.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910082 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837953 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179505 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036764 | BC036171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036764 | BC022765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036764 | BC024510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036764 | AK090284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036764 | BC034192.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | BC036171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | BC022765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | BC024510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | AK090284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | BC034192.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | BC036171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | BC022765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | BC024510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | AK090284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P435561 | BC034192.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28047592 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07454256 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179505 | - | - |
| Whole Mouse Genome G4122A | A_52_P435561 | BC036171.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P435561 | BC022765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P435561 | BC024510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P435561 | AK090284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P435561 | BC034192.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P435561 | BC036171.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P435561 | BC022765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P435561 | BC024510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P435561 | AK090284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P435561 | BC034192.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P418644 | AK087566.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127566 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116068 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09131600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09305137 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09131604 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09131605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28047657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09131600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09131608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09365186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09539528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09257435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08982285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09365182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09539888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09131602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28047635 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380713 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380714 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433870_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380712 | - | - |
| Mouse Genome 430 2.0 Array | 1433870_at | - | - |
| Mouse Genome 430 2.0 Array | 1433870_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217138_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217138_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100014023.mm.1 | NM_146026.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014023.mm.1 | NM_146026.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014023.mm.1 | TC1100001524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014029.mm.1 | NM_146026.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014029.mm.1 | NM_146026.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014029.mm.1 | TC1100001524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014024.mm.1 | NM_146026.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014024.mm.1 | NM_146026.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014024.mm.1 | TC1100001524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014027.mm.1 | NM_146026.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014027.mm.1 | NM_146026.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014027.mm.1 | TC1100001524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014030.mm.1 | NM_146026.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014030.mm.1 | NM_146026.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014030.mm.1 | TC1100001524.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433870_PM_at | - | - |
| Mouse Expression Set 430 | 1433870_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433870_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433870_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104480341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104480341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 110010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000110010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246558 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215227 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246558 | - | - |
| mouse-6 v1.1 (Illumina) | scl00217138.2_80-S | - | - |
| mouse-6 v1.1 (Illumina) | scl40972.4_107-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104480341 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 110010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00217138.2_80-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104480341 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 110010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2788191 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1246558 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2788191 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215227 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246558 | - | - |

Prr15l
Genomic Map