Assay Details
| Gene Symbol: Soat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146064.1 | NP_666176.1 | 7 - 8 | 851 | 72 | |
| GenBank mRNA | BC025931.1 | - | 7 - 8 | 851 | 5099063 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Soat2
Genomic Map