Assay Details
| Gene Symbol: Lemd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146075.2 | NP_666187.2 | 2 - 3 | 826 | 58 | |
| GenBank mRNA | AK082131.1 | - | 2 - 3 | 633 | 58 | |
| AK143224.1 | - | 2 - 3 | 811 | 58 | ||
| AY640307.1 | - | 2 - 3 | 806 | 58 | ||
| BC026588.1 | - | 2 - 3 | 136 | 4972900 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lemd2
Genomic Map