Assay Details
| Gene Symbol: Gsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206367.1 | NP_001193296.1 | 17 - 18 | 2238 | 73 | |
| NM_001206368.1 | NP_001193297.1 | 16 - 17 | 2154 | 73 | ||
| NM_001206369.1 | NP_001193298.1 | 16 - 17 | 2199 | 73 | ||
| NM_146120.4 | NP_666232.2 | 16 - 17 | 2261 | 73 | ||
| GenBank mRNA | AK076156.1 | - | 16 - 17 | 2201 | 73 | |
| AK089934.1 | - | 17 - 18 | 2193 | 73 | ||
| AK135632.1 | - | 16 - 17 | 1974 | 73 | ||
| AK143664.1 | - | 16 - 17 | 2228 | 73 | ||
| AK146969.1 | - | 17 - 18 | 2155 | 73 | ||
| AK148957.1 | - | 17 - 18 | 2218 | 73 | ||
| AK150359.1 | - | 17 - 18 | 2161 | 73 | ||
| AK151684.1 | - | 17 - 18 | 2180 | 73 | ||
| AK153029.1 | - | 17 - 18 | 2158 | 73 | ||
| AK166831.1 | - | 16 - 17 | 2174 | 73 | ||
| AK166932.1 | - | 17 - 18 | 2152 | 73 | ||
| AK167918.1 | - | 17 - 18 | 2168 | 73 | ||
| AK168461.1 | - | 17 - 18 | 2171 | 73 | ||
| AK168532.1 | - | 17 - 18 | 2172 | 73 | ||
| AK168707.1 | - | 16 - 17 | 2107 | 73 | ||
| AK168725.1 | - | 17 - 18 | 2155 | 73 | ||
| AK168829.1 | - | 16 - 17 | 2207 | 73 | ||
| AK169041.1 | - | 17 - 18 | 2158 | 73 | ||
| AK169670.1 | - | 17 - 18 | 2183 | 73 | ||
| AK213866.1 | - | 3 - 4 | 276 | 73 | ||
| AK217606.1 | - | 2 - 3 | 93 | 73 | ||
| AK218612.1 | - | 2 - 3 | 93 | 73 | ||
| AK219663.1 | - | 2 - 3 | 93 | 73 | ||
| BC023143.1 | - | 16 - 17 | 2199 | 3711602 | 73 | |
| BC060377.1 | - | 17 - 18 | 2186 | 4983664 | 73 | |
More Information
Set Membership: |
|

Gsn
Genomic Map