Assay Details
| Gene Symbol: Pcif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146129.3 | NP_666241.1 | 3 - 4 | 435 | 71 | |
| GenBank mRNA | AK089358.1 | - | 3 - 4 | 436 | 71 | |
| AK150740.1 | - | 2 - 3 | 326 | 71 | ||
| AK211200.1 | - | 2 - 3 | 343 | 71 | ||
| BC031431.1 | - | 3 - 4 | 404 | 4241283 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcif1
Genomic Map