Assay Details
| Gene Symbol: Usp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146144.4 | NP_666256.2 | 1 - 2 | 434 | 112 | |
| GenBank mRNA | AK080882.1 | - | 1 - 2 | 435 | 112 | |
| BC018179.1 | - | 1 - 2 | 314 | 3585191 | 112 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK080882.1,BC018179.1,NM_146144.4 transcripts.
Set Membership: |
|

Usp1
Genomic Map