Assay Details
| Gene Symbol: Usp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301414.1 | NP_001288343.1 | 7 - 8 | 1926 | 83 | |
| NM_146144.4 | NP_666256.2 | 7 - 8 | 1907 | 83 | ||
| GenBank mRNA | AK080882.1 | - | 7 - 8 | 1908 | 83 | |
| BC018179.1 | - | 7 - 8 | 1787 | 3585191 | 83 | |
| BC020007.1 | - | 7 - 8 | 1759 | 3980469 | 83 | |
| BC023812.1 | - | 4 - 5 | 1081 | 83 | ||
More Information
Set Membership: |
|

Usp1
Genomic Map