Assay Details
| Gene Symbol: Arhgap24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286468.1 | NP_001273397.1 | 3 - 4 | 482 | 67 | |
| NM_029270.2 | NP_083546.2 | 5 - 6 | 1084 | 67 | ||
| NM_146161.3 | NP_666273.1 | 4 - 5 | 890 | 67 | ||
| GenBank mRNA | AK039617.1 | - | 2 - 3 | 1373 | 67 | |
| AK080986.1 | - | 5 - 6 | 1084 | 67 | ||
| AK134817.1 | - | 3 - 4 | 708 | 67 | ||
| BC023344.1 | - | 3 - 4 | 496 | 5064885 | 67 | |
| BC025502.1 | - | 4 - 5 | 739 | 5321985 | 67 | |
| BC027070.1 | - | 2 - 3 | 328 | 5358648 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap24
Genomic Map