Assay Details
| Gene Symbol: Cnot3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146176.3 | NP_666288.1 | 9 - 10 | 1199 | 59 | |
| GenBank mRNA | AK141718.1 | - | 9 - 10 | 1179 | 59 | |
| AK173003.1 | - | 5 - 6 | 613 | 59 | ||
| BC030332.1 | - | 9 - 10 | 1128 | 5400871 | 59 | |
| BC053437.1 | - | 9 - 10 | 960 | 6394085 | 59 | |
| BC141041.1 | - | 9 - 10 | 999 | 9055653 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot3
Genomic Map