Assay Details
| Gene Symbol: Rps6kl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146244.4 | NP_666356.2 | 3 - 4 | 515 | 90 | |
| GenBank mRNA | AK031847.1 | - | 3 - 4 | 506 | 90 | |
| AK044050.1 | - | 3 - 4 | 506 | 90 | ||
| BC024798.1 | - | 3 - 4 | 457 | 5358742 | 90 | |
| BC027298.1 | - | 3 - 4 | 469 | 3672127 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kl1
Genomic Map