Assay Details
| Gene Symbol: Rps6kl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031847.1 | - | 7 - 8 | 1371 | 71 | |
| AK036734.1 | - | 1 - 2 | 915 | 71 | ||
| BC024798.1 | - | 6 - 7 | 1274 | 5358742 | 71 | |
More Information
Set Membership: |
|

Rps6kl1
Genomic Map