Assay Details
| Gene Symbol: Prdx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007452.2 | NP_031478.1 | 1 - 2 | 108 | 61 | |
| GenBank mRNA | AK002448.1 | - | 1 - 2 | 42 | 61 | |
| BC005626.1 | - | 1 - 2 | 59 | 3963113 | 61 | |
| BY318475.1 | - | 1 - 2 | 111 | 61 | ||
| M28723.1 | - | 1 - 2 | 84 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prdx3
Genomic Map