Assay Details
| Gene Symbol: Stard3nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002760.1 | - | 3 - 4 | 469 | 64 | |
| AK018331.1 | - | 4 - 5 | 470 | 64 | ||
| AK152116.1 | - | 4 - 5 | 532 | 64 | ||
| AK152378.1 | - | 4 - 5 | 483 | 64 | ||
| BC003334.1 | - | 4 - 5 | 486 | 3256725 | 64 | |
| BG084215.2 | - | 4 - 5 | 488 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard3nl
Genomic Map