Assay Details
| Gene Symbol: Dyx1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163725.1 | NP_001157197.1 | 4 - 5 | 619 | 70 | |
| NM_026314.3 | NP_080590.3 | 6 - 7 | 979 | 70 | ||
| GenBank mRNA | AK005832.1 | - | 3 - 4 | 429 | 70 | |
| AK015345.1 | - | 2 - 3 | 158 | 70 | ||
| AY428507.1 | - | 6 - 7 | 979 | 70 | ||
| AY464142.1 | - | 4 - 5 | 619 | 70 | ||
| BC026462.1 | - | 6 - 7 | 936 | 4483015 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dyx1c1
Genomic Map