Assay Details
| Gene Symbol: Ppm1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026447.4 | NP_080723.3 | 5 - 6 | 843 | 80 | |
| NM_198931.3 | NP_945149.2 | 5 - 6 | 847 | 80 | ||
| GenBank mRNA | AB474373.1 | - | 5 - 6 | 882 | 80 | |
| AK013149.1 | - | 5 - 6 | 776 | 80 | ||
| AK046387.1 | - | 742 | 80 | |||
| AK087999.1 | - | 5 - 6 | 881 | 80 | ||
| AY332616.1 | - | 5 - 6 | 850 | 80 | ||
| BC058248.1 | - | 5 - 6 | 769 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppm1m
Genomic Map