Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 18 - 19 | 1812 | 68 | |
| GenBank mRNA | AK038318.1 | - | 18 - 19 | 1790 | 68 | |
| AK043764.1 | - | 7 - 8 | 657 | 68 | ||
| AK122320.1 | - | 17 - 18 | 1818 | 68 | ||
| AK145716.1 | - | 18 - 19 | 1813 | 68 | ||
| AK161181.1 | - | 18 - 19 | 1762 | 68 | ||
| AK163765.1 | - | 17 - 18 | 1800 | 68 | ||
| AK168388.1 | - | 18 - 19 | 1799 | 68 | ||
| BC032177.1 | - | 16 - 17 | 2039 | 68 | ||
| BC040393.1 | - | 18 - 19 | 1736 | 68 | ||
| BC049979.1 | - | 18 - 19 | 1788 | 68 | ||
| BC056942.1 | - | 18 - 19 | 1739 | 6827177 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam21
Genomic Map