Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 23 - 24 | 2563 | 84 | |
| GenBank mRNA | AK038318.1 | - | 23 - 24 | 2541 | 84 | |
| AK043764.1 | - | 12 - 13 | 1408 | 84 | ||
| AK122320.1 | - | 22 - 23 | 2569 | 84 | ||
| AK145716.1 | - | 23 - 24 | 2564 | 84 | ||
| AK161181.1 | - | 23 - 24 | 2513 | 84 | ||
| AK163765.1 | - | 22 - 23 | 2551 | 84 | ||
| AK168388.1 | - | 23 - 24 | 2552 | 84 | ||
| BC032177.1 | - | 21 - 22 | 2789 | 84 | ||
| BC040393.1 | - | 23 - 24 | 2487 | 84 | ||
| BC049979.1 | - | 23 - 24 | 2539 | 84 | ||
| BC056942.1 | - | 23 - 24 | 2490 | 6827177 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam21
Genomic Map