Assay Details
| Gene Symbol: Dcaf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026893.3 | NP_081169.2 | 4 - 5 | 939 | 76 | |
| GenBank mRNA | AK005099.1 | - | 4 - 5 | 885 | 76 | |
| AK030173.1 | - | 4 - 5 | 891 | 76 | ||
| AK031089.1 | - | 4 - 5 | 869 | 76 | ||
| AK078036.1 | - | 2 - 3 | 240 | 76 | ||
| AK087517.1 | - | 4 - 5 | 823 | 76 | ||
| AK090028.1 | - | 4 - 5 | 888 | 76 | ||
| AK143635.1 | - | 4 - 5 | 892 | 76 | ||
| AK173289.1 | - | 4 - 5 | 799 | 76 | ||
| BC023852.1 | - | 4 - 5 | 875 | 5346910 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf12
Genomic Map