Assay Details
| Gene Symbol: H1fnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027304.2 | NP_081580.2 | 1 | 979 | 57 | |
| GenBank mRNA | AB016273.1 | - | 1 | 951 | 57 | |
| AY496853.1 | - | 1 | 977 | 57 | ||
| BC052363.1 | - | 1 | 965 | 6773647 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30473863 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30473865 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30473864 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30473862 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | BC052363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | NM_027304.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | AB016273.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | AK006402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | AY496853.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | BC052363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | NM_027304.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | AB016273.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | AK006402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P250001 | AY496853.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104659 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142547 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | BC052363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | NM_027304.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | AB016273.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | AK006402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | AY496853.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | BC052363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | NM_027304.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | AB016273.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | AK006402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P250001 | AY496853.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30473883 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104659 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142547 | - | - |
| Whole Mouse Genome G4122A | A_51_P250001 | BC052363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P250001 | NM_027304.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P250001 | AB016273.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P250001 | AK006402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P250001 | AY496853.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P250001 | BC052363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P250001 | NM_027304.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P250001 | AB016273.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P250001 | AK006402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P250001 | AY496853.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114036 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142547 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104659 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12446811 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12446807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12446812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12446813 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12446817 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30473864 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30473865 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30473863 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30473862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30473905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12446813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11945383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11864310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30473887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12053149 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10432137 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432136 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432138 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430181_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441954_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432135 | - | - |
| Mouse Genome 430 2.0 Array | 1441954_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1441954_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1430181_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430181_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70069_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70069_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500017614.mm.1 | NM_027304.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017614.mm.1 | NM_027304.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017614.mm.1 | TC1500002169.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430181_PM_at | - | - |
| Mouse Expression Set 430 | 1430181_at | - | - |
| Mouse Expression Set 430 | 1441954_s_at | - | - |
| Mouse Expression Set 430 | 1441954_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430181_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441954_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430181_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441954_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 845799 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480070 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480070 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664702 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664702 | - | - |
| mouse-6 v1.1 (Illumina) | scl46773.1.15_167-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480070 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46773.1.15_167-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480070 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2664702 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2664702 | - | - |

H1fnt
Genomic Map