Assay Details
| Gene Symbol: Zdhhc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027476.3 | NP_081752.2 | 1 - 2 | 361 | 87 | |
| GenBank mRNA | AK017737.1 | - | 1 - 2 | 326 | 87 | |
| AK043923.1 | - | 1 - 2 | 318 | 87 | ||
| AK169337.1 | - | 1 - 2 | 320 | 87 | ||
| AK170233.1 | - | 1 - 2 | 345 | 87 | ||
| AK170706.1 | - | 1 - 2 | 345 | 87 | ||
| AK171568.1 | - | 1 - 2 | 345 | 87 | ||
| BC071194.1 | - | 1 - 2 | 302 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31925605 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31925604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31925606 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | AK172151.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | BC056621.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | BC071194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | AK170233.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | AK170706.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | AK172151.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | BC056621.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | BC071194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | AK170233.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208870 | AK170706.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686675 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909872 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391434 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | AK172151.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | BC056621.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | BC071194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | AK170233.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | AK170706.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | AK172151.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | BC056621.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | BC071194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | AK170233.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208870 | AK170706.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31925622 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09091852 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03542004 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03542009 | - | - |
| Whole Mouse Genome G4122A | A_52_P537140 | AK047235.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | AK172151.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | BC056621.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | BC071194.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | AK170233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | AK170706.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | AK172151.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | BC056621.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | BC071194.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | AK170233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208870 | AK170706.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P237352 | BC056621.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P237352 | BC071194.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P237352 | AK170706.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P237352 | AK171568.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P237352 | AK043923.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124667 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09091852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14367001 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31925604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31925605 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31925606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31925707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14232439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14273479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14279823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14257797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14273469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14232443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14232427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14257796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14366997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31925633 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460487 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460488 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460489 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452719_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428278_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460485 | - | - |
| Mouse Genome 430 2.0 Array | 1428278_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452719_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428278_at | - | - |
| Mouse Genome 430 2.0 Array | 1452719_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70605_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70605_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900000660.mm.1 | NM_027476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000660.mm.1 | NM_027476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000660.mm.1 | TC1900000047.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000659.mm.1 | NM_027476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000659.mm.1 | NM_027476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000659.mm.1 | TC1900000047.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452719_PM_at | - | - |
| Mouse Expression Set 430 | 1428278_at | - | - |
| Mouse Expression Set 430 | 1452719_at | - | - |
| Mouse Expression Set 430 | 1428278_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452719_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428278_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452719_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428278_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 751277 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 101690671 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5700487 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2060400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005700487 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251104 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697127 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251104 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2697127 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2567347 | - | - |
| mouse-6 v1.1 (Illumina) | scl070605.1_38-S | - | - |
| mouse-6 v1.1 (Illumina) | scl52824.3_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060400 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5700487 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101690671 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52824.3_17-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl070605.1_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060400 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700487 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101690671 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1251104 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2697127 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2860580 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251104 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2567347 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2697127 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2860580 | - | - |

Zdhhc24
Genomic Map