Assay Details
| Gene Symbol: Noxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027988.4 | NP_082264.2 | 2 - 3 | 1085 | 59 | |
| GenBank mRNA | AF399754.1 | - | 1 - 2 | 71 | 59 | |
| AF539797.1 | - | 1 - 2 | 71 | 59 | ||
| AK009605.1 | - | 1 - 2 | 180 | 59 | ||
| AK088226.1 | - | 2 - 3 | 1044 | 59 | ||
| BC019525.1 | - | 1 - 2 | 97 | 4220018 | 59 | |
| BC119527.1 | - | 1 - 2 | 173 | 59 | ||
| BC119528.1 | - | 1 - 2 | 173 | 59 | ||
| BC127235.1 | - | 1 - 2 | 173 | 59 | ||
| BC127236.1 | - | 1 - 2 | 173 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111907 | AK137554.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111907 | BC019525.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111907 | AK009605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111907 | AK088226.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111907 | AF539797.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604132 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140796 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111907 | AK137554.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111907 | BC019525.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111907 | AK009605.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111907 | AK088226.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111907 | AF539797.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625940 | AK137554.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625940 | AK157993.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625940 | AK009605.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625940 | AK088226.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625940 | NM_027988.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099081 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140796 | - | - |
| Whole Mouse Genome G4122A | A_52_P625940 | AK137554.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625940 | AK157993.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625940 | AK009605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625940 | AK088226.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625940 | NM_027988.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116755 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140796 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13380576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13380618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13380625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13312253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13380615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13312252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099071 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442575 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442573 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442577 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442578 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442569 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425151_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442565 | - | - |
| Mouse Genome 430 2.0 Array | 1425151_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425151_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71893_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71893_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700002663.mm.1 | NM_027988.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002663.mm.1 | NM_027988.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002663.mm.1 | TC1700000348.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002667.mm.1 | NM_027988.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002667.mm.1 | NM_027988.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002667.mm.1 | TC1700000348.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002665.mm.1 | NM_027988.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002665.mm.1 | NM_027988.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002665.mm.1 | TC1700000348.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002669.mm.1 | NM_027988.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002669.mm.1 | NM_027988.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002669.mm.1 | TC1700000348.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002658.mm.1 | NM_027988.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002658.mm.1 | NM_027988.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002658.mm.1 | TC1700000348.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425151_PM_a_at | - | - |
| Mouse Expression Set 430 | 1425151_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425151_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425151_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425151_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425151_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7040075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4050731 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7040075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050731 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007040075 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2683856 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240610 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240610 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2683856 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001724.1_23-S | - | - |
| mouse-6 v1.1 (Illumina) | scl51008.5.1_229-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7040075 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050731 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001724.1_23-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51008.5.1_229-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040075 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050731 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240610 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2683856 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240610 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2683856 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001724.1_23-S | - | - |

Noxo1
Genomic Map